Order Entry
Belgium
ContactUsLinkComponent
Cetyldimethylethylammonium bromide, high purity
Irritant
Cetyldimethylethylammonium bromide, high purity
Catalog # SPCME1054-12KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Cetyldimethylethylammonium bromide, high purity
Catalog # SPCME1054-12KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specification Test Results

About this item

 :
 :378.4741 g/mol
Storage Temperature:Ambient
EINECS:204-672-8
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:124-03-8
Formula:C₂₀H₄₄BrN
MDL Number:MFCD00011774