Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bismuth(III) nitrate basic, powder USP

Oxidizing
Irritant
Supplier:   Spectrum Chemical
Bismuth(III) nitrate basic, powder USP
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents

Form: Powder

Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Density:4.928 g/cm³ (20 °C)
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Specifications for Product Family

Specification Test Results

  • Assay:
    79% Min.
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 12 kg
    Log in for availability
    Log in for availability
    1 * 12 kg
    1 * 12 kg - 3 800,00 €
    3 800,00 €
    1 * 12 kg
       3 800,00 €

  • 50 kg
    Log in for availability
    Log in for availability
    1 * 50 kg
    1 * 50 kg - 7 040,00 €
    7 040,00 €
    1 * 50 kg
       7 040,00 €