Order Entry
Belgium
Orders LinkContactUsLinkComponent
1,4-Dimethoxybenzene

1,4-Dimethoxybenzene

Drum, 25 kg
Catalog # SPCMD2455-25KGBL
Supplier:   Spectrum Chemical
1,4-Dimethoxybenzene
Catalog # SPCMD2455-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  150-78-7
1,4-Dimethoxybenzene

Drum, 25 kg

CAS Number:  150-78-7
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  p-Dimethoxybenzene, Hydroquinone dimethyl ether
  • CAS Number:150-78-7
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00008401
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:212.6 °C (1013 hPa)
  • Melting Pt:54…56 °C
  • Density:0.874 g/cm³ (24.9 °C)
  • Storage Temperature:Ambient
  • EINECS:205-771-9
Specifications

Specification Test Results

  • Assay:
    98% Min.

Specifications

  • Pack type:
    Drum
  • Pk:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.