2-Phénylethanol USP
Supplier: Spectrum Chemical
Jump to:
About this item
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:432 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1,02 g/cm³ (20 °C)
- Flash Pt:102 °C
- Storage Temperature:Température ambiante
- EINECS:200-456-2
- UN:2810
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:432 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1,02 g/cm³ (20 °C)
- Flash Pt:102 °C
- Storage Temperature:Température ambiante
- EINECS:200-456-2
- UN:2810
- ADR:6.1,III
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



