ContactUsLinkComponent
Test strips, zinc, MQuant®, Supelco®
Test strips, zinc, MQuant®, Supelco®
Catalog # 1.17953.0001
Supplier:  Merck
CAS Number:  
undefined
Test strips, zinc, MQuant®, Supelco®
Catalog # 1.17953.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Dithizone
  • Pk:
    100 Tests
  • Description:
    Zinc test
  • Environmentally Preferable:
  • Type:
    MQ
  • Range:
    4-10-20-50 mg/l Zn
Specifications
About this item
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: Just dip, wait and measure visually against colour scale
  • Safe: No auxiliary reagents necessary for most kits
  • Immediate results within a few minutes enables fast decisions
  • Portable: Can be used inside or outside the laboratory