1-(4-Methoxyphenyl)ethanol 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3319-15-1
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00016857
:152.19 g/mol
Boiling Pt:94…96 °C (1 mmHg)
Density:1.079
Flash Pt:74 °C (165 °F)
Storage Temperature:Ambient
EINECS:222-019-5
Related Information
Frequently Bought Together