Silver 0.02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 10 nm (Abs. 390-400 nm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-22-4
:
Formula:Ag
MDL Number:MFCD00003397
:107.87 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together