Cilostamide
10 mg
CAS No.: 68550-75-4 Molecular Formula: C20H26N2O3 Formula Weight: 342.43 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00673958
Applications: A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase
: N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide
- :68550-75-4
- :C₂₀H₂₆N₂O₃
- :MFCD00673958
- :342.43 g/mol
- Pk:10 mg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



