Order Entry
Belgium
Orders LinkContactUsLinkComponent
Factor XA protease digestion buffer

Factor XA protease digestion buffer

Factor XA Protease Digestion Buffer, 250 mL
Catalog # J63878.AK
Factor XA protease digestion buffer
Catalog # J63878.AK
CAS Number:  
Factor XA protease digestion buffer
Factor XA protease digestion buffer

Factor XA Protease Digestion Buffer, 250 mL

CAS Number:  
Price
68,30 €
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid
Applications: For His-tag protein purification from mammalian cells

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Factor XA Protease Digestion Buffer
  • Cat. No.:
    J63878.AK
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.