Order Entry
Belgium
Orders LinkContactUsLinkComponent
TEA, 0,2 M buffer soln., pH 8,0

TEA, 0,2 M buffer soln., pH 8,0

TEA buffer solution, 0,2 M, 8,0, 250 mL
Catalog # J63793.AK
TEA, 0,2 M buffer soln., pH 8,0
Catalog # J63793.AK
CAS Number:  
TEA, 0,2 M buffer soln., pH 8,0
TEA, 0,2 M buffer soln., pH 8,0

TEA buffer solution, 0,2 M, 8,0, 250 mL

CAS Number:  
Price
112,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 102-71-6
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002855

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    250 mL
  • Description:
    TEA buffer solution
  • pH value:
    8,0
  • Cat. No.:
    J63793.AK
  • Odour/scent:
    Odourless
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00002855
  • Appearance:
    Colourless
  • CAS No.:
    102-71-6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.