IEF Anode buffer (50X)
Supplier: Thermo Fisher Scientific
About this item
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD03846400
Applications: For isoelectric focusing (IEF)
2 Options Available Below
Product Details & Documents
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD03846400
Applications: For isoelectric focusing (IEF)
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J63303.AP
- J63303.K2
- Appearance:
- Colourless
- Colourless
- Storage temperature:
- 2°C to 8°C
- 2°C to 8°C
- Physical form:
- Liquid
- Liquid
- MDL No.:
- MFCD03846400
- MFCD03846400
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- J63303.APIEF Anode buffer (50X)Specifications:
More
Specifications
Cat. No.J63303.APAppearanceColourlessStorage temperature2°C to 8°CPhysical formLiquidMDL No.MFCD03846400 - J63303.K2IEF Anode buffer (50X)Specifications:
More
Specifications
Cat. No.J63303.K2AppearanceColourlessStorage temperature2°C to 8°CPhysical formLiquidMDL No.MFCD03846400
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



