Order Entry
Belgium
ContactUsLinkComponent
HEPES protein extraction buffer (5X)
HEPES protein extraction buffer (5X)
Catalog # J63297.AK
CAS Number:  
HEPES protein extraction buffer (5X)
Catalog # J63297.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

Physical Form: Liquid
Appearance: Colourless