Order Entry
Belgium
Orders LinkContactUsLinkComponent
Transformation buffer-2, pH 7,5

Transformation buffer-2, pH 7,5

Transformation buffer, 7,5, 250 mL
Catalog # J63293.AK
Transformation buffer-2, pH 7,5
Catalog # J63293.AK
CAS Number:  
Transformation buffer-2, pH 7,5
Transformation buffer-2, pH 7,5

Transformation buffer, 7,5, 250 mL

CAS Number:  
Price
152,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Applications: For transformation of competent cells

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Transformation buffer
  • pH value:
    7,5
  • Cat. No.:
    J63293.AK
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid

Specifications