Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

TAPS, 0,2 M buffer soln., pH 8,5

TAPS buffer soln, 0,2 M, 8,5, 100 mL
Catalog # J63268.AE
TAPS, 0,2 M buffer soln., pH 8,5
Catalog # J63268.AE
CAS Number:  
TAPS, 0,2 M buffer soln., pH 8,5
TAPS, 0,2 M buffer soln., pH 8,5

TAPS buffer soln, 0,2 M, 8,5, 100 mL

CAS Number:  
Price
96,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    100 mL
  • Description:
    TAPS buffer soln
  • pH value:
    8,5
  • Cat. No.:
    J63268.AE
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00069903
  • CAS No.:
    91000-53-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.