Order Entry
Belgium
ContactUsLinkComponent
Saponin permeating solution, 0,5% w/v solution in PBS (10X)
Saponin permeating solution, 0,5% w/v solution in PBS (10X)
Catalog # J63209.AK
CAS Number:  
Saponin permeating solution, 0,5% w/v solution in PBS (10X)
Catalog # J63209.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Saponin Permeating Solution
  • Storage conditions:
    RT
  • Form:
    Liquid
  • Solubility:
    Difficult to Mix

Specifications

About this item

Saponin permeating solution, is used as a cell permeabilising agent.

Physical Form: Liquid

Applications: Cell permeabilizing agent