Order Entry
Belgium
Orders LinkContactUsLinkComponent
Alkaline running buffer solution

Alkaline running buffer solution

Alkaline running buffer solution, 500 mL
Catalog # J63170.AP
Alkaline running buffer solution
Catalog # J63170.AP
CAS Number:  
Alkaline running buffer solution
Alkaline running buffer solution

Alkaline running buffer solution, 500 mL

CAS Number:  
Price
61,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

Physical Form: Liquid
UN No.:UN1824

Contains 0.5 M NaOH and 50 mM EDTA

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Alkaline running buffer solution
  • Cat. No.:
    J63170.AP
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.