Order Entry
Belgium
ContactUsLinkComponent
Enzyme storage buffer in PBS and glycerol, autoclaved
Enzyme storage buffer in PBS and glycerol, autoclaved
Catalog # J63115.AK
CAS Number:  
Enzyme storage buffer in PBS and glycerol, autoclaved
Catalog # J63115.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Enzyme Storage Buffer in PBS and Glycerol
  • Physical form:
    Liquid

Specifications

About this item

Physical Form:
Applications: For frozen storage of enzymes.