Order Entry
Belgium
ContactUsLinkComponent
 
Buffer kit for antibody purification on protein A column
  J63014.QA
 :  
Buffer kit for antibody purification on protein A column
  J63014.QA
 :  

 

  • Pk:
    1 KIT
  • Description:
    Buffer kit for antibody purification
  • Physical form:
    Liquid

 

 

Physical Form: Liquid

Applications: Buffers to be used when performing antibody purification using immobilized Protein A.

 : This kit contains: 1 litre of 1 M Tris-HCl (pH 8.0) and 500 ml of 100 mM glycine (pH 3.0).