Order Entry
Belgium
ContactUsLinkComponent
TRIS-glycine-native running buffer (10x), pH 8,5
TRIS-glycine-native running buffer (10x), pH 8,5
Certificates
TRIS-glycine-native running buffer (10x), pH 8,5
Certificates

Specifications

  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together