Order Entry
Belgium
ContactUsLinkComponent
Phosphate-buffered saline (PBS, 10X), RNase free
Phosphate-buffered saline (PBS, 10X), RNase free
Certificates
Phosphate-buffered saline (PBS, 10X), RNase free
Certificates

Specifications

  • MDL No.:
    MFCD00131855
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together