HEPES-buffered saline, pH 7,0 (2X for transfection)
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
CAS No.: 7365-45-9
Physical Form: Liquid
MDL No.: MFCD00006158
Spécifications
- CAS No.:7365-45-9
- MDL No.:MFCD00006158
- Physical form:Liquid
Spécifications
Frequently Bought Together