Order Entry
Belgium
Orders LinkContactUsLinkComponent
 
GTE buffer
Certificates
GTE buffer

2 Options Available Below

Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Applications: For miniprep and DNA purification

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J62597.AE
  • J62597.AK
  • Physical form:
  • Liquid
  • Liquid
  • Storage temperature:
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 41,90 €
    41,90 €
    1 * 100 mL
       41,90 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62597.AE
    Physical form
    Liquid
    Storage temperature
    2°C to 8°C
  • 250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 63,80 €
    63,80 €
    1 * 250 mL
       63,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62597.AK
    Physical form
    Liquid
    Storage temperature
    2°C to 8°C