Order Entry
Belgium
Orders LinkContactUsLinkComponent
TE buffer, pH 7,6, RNAse free

TE buffer, pH 7,6, RNAse free

TE buffer, pH 7,6, RNAse free
Certificates
TE buffer, pH 7,6, RNAse free

2 Options Available Below

Product Details & Documents

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. TE buffer functions to solubilise DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

Physical Form: Liquid
MDL No.: MFCD00236359

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J62285.XE
  • J62285.AK
  • Physical form:
  • Liquid
  • Liquid
  • MDL No.:
  • MFCD00236359
  • MFCD00236359
  • Storage temperature:
  • Ambient temperature
  • Ambient temperature

Specifications

Product Family Options

Product Information

  • pH value
    Pk
    Availability
    Price
  • J62285.XE
    TE buffer, RNAse free
    TE buffer, RNAse free
    7,6
    125 mL
    Log in for availability
    Log in for availability
    1 * 125 mL
    1 * 125 mL - 67,20 €
    67,20 €
    1 * 125 mL
       67,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62285.XE
    Physical form
    Liquid
    MDL No.
    MFCD00236359
    Storage temperature
    Ambient temperature
  • J62285.AK
    TE buffer, RNAse free
    TE buffer, RNAse free
    7,6
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 119,00 €
    119,00 €
    1 * 250 mL
       119,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62285.AK
    Physical form
    Liquid
    MDL No.
    MFCD00236359
    Storage temperature
    Ambient temperature
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.