Order Entry
Belgium
ContactUsLinkComponent
IEF Cathode buffer (10X) with lysine and arginine
IEF Cathode buffer (10X) with lysine and arginine
Catalog # J62204.AP
CAS Number:  
IEF Cathode buffer (10X) with lysine and arginine
Catalog # J62204.AP
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    IEF Cathode buffer (10X) with lysine and arginine
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

Physical Form: Liquid
Appearance: Colourless
Applications: For isoelectric focusing