Order Entry
Belgium
ContactUsLinkComponent
RBC lysis buffer for mouse
RBC lysis buffer for mouse
Certificates
RBC lysis buffer for mouse
Certificates

Specifications

  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Related Information

Frequently Bought Together