AMP, 0.2 M Buffer Solutions
AMP buffer solution, 0,2 M, 9,5, Glass bottle, 100 mL
2-Amino-2-methyl-1-propanol buffer, colorless and odorless.
CAS No.: 124-68-5
MDL No.: MFCD00008051
: Storage Temperature: 2 to 8 °C
- Concentration:0,2 M
- Pack type:Glass bottle
- Pk:100 mL
- Description:AMP buffer solution
- pH value:9,5
- Cat. No.:J63988.AE
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00008051
- Appearance:Colourless
- CAS No.:124-68-5



