Order Entry
Belgium
ContactUsLinkComponent
 
Ciprofloxacin hydrochloride monohydrate 98.0-102.0%
 
Ciprofloxacin hydrochloride monohydrate 98.0-102.0%
 

 

  • Appearance (Color):
    White to off-white
  • Form:
    Lyophilised powder
  • Assay (unspecified):
    98,0 - 102,0%
  • Residue on ignition:
    0,1% max.
  • Impurity content:
    0,002% max. (Heavy metals)
  • Water Content (Karl Fischer Titration):
    4,7 - 6,7%
  • Solubility:
    Soluble in Water (35 mg/ml)

 

Frequently Bought Together