Order Entry
Belgium
ContactUsLinkComponent
 
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
 
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula
 

 

  • MDL No.:
    MFCD00131855
  • Physical form:
    Liquid
  • Appearance:
    Colourless

 

Frequently Bought Together