Orders LinkContactUsLinkComponent
LDS-sample buffer (4X), non-reducing
LDS-sample buffer (4X), non-reducing
LDS-sample buffer (4X), non-reducing
LDS-sample buffer (4X), non-reducing
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.
Physical Form: Liquid
Appearance: Blue
This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.
  • Catalog No:
  • J61894.AC
  • J61894.AD
  • Appearance:
  • Blue
  • Blue
  • Physical form:
  • Liquid
  • Liquid
Product Family Options
  • Description
    Pk
  • LDS-Sample Buffer (4X), non-reducing
    25 mL
    1 * 25 mL
    1 * 25 mL - 87,70 €
    87,70 €
    1 * 25 mL
       87,70 €
    Specifications
    Cat. No.
    J61894.AC
    Appearance
    Blue
    Physical form
    Liquid
  • LDS-Sample Buffer (4X), non-reducing
    50 mL
    1 * 50 mL
    1 * 50 mL - 144,00 €
    144,00 €
    1 * 50 mL
       144,00 €
    Specifications
    Cat. No.
    J61894.AD
    Appearance
    Blue
    Physical form
    Liquid