TES, 0,2 M buffer soln., pH 7,5
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 70331-82-7
Physical Form: Liquid
MDL No.: MFCD00065482
2 Options Available Below
Product Details & Documents
CAS No.: 70331-82-7
Physical Form: Liquid
MDL No.: MFCD00065482
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J61350.AE
- J61350.AK
- Physical form:
- Liquid
- Liquid
- CAS No.:
- 70331-82-7
- 70331-82-7
- MDL No.:
- MFCD00065482
- MFCD00065482
Specifications
Product Family Options
Product Information
- ConcentrationpH valuePkAvailabilityPrice
- J61350.AETES buffer solutionSpecifications:
More
Specifications
Cat. No.J61350.AEPhysical formLiquidCAS No.70331-82-7MDL No.MFCD00065482 - J61350.AKTES buffer solutionSpecifications:
More
Specifications
Cat. No.J61350.AKPhysical formLiquidCAS No.70331-82-7MDL No.MFCD00065482



