Order Entry
Belgium
Orders LinkContactUsLinkComponent
Laemmli SDS sample buffer, reducing (6X)

Laemmli SDS sample buffer, reducing (6X)

Laemmli SDS sample buffer, reducing (6X)
Certificates
Laemmli SDS sample buffer, reducing (6X)

2 Options Available Below

Product Details & Documents

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J61337.AC
  • J61337.AD
  • Appearance:
  • Blue
  • Blue
  • Physical form:
  • Liquid
  • Liquid

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • J61337.AC
    Laemmli SDS Sample Buffer, reducing, 6X
    Laemmli SDS Sample Buffer, reducing, 6X
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 105,00 €
    105,00 €
    1 * 25 mL
       105,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61337.AC
    Appearance
    Blue
    Physical form
    Liquid
  • J61337.AD
    Laemmli SDS Sample Buffer, reducing, 6X
    Laemmli SDS Sample Buffer, reducing, 6X
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 165,00 €
    165,00 €
    1 * 50 mL
       165,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61337.AD
    Appearance
    Blue
    Physical form
    Liquid
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.