Glucose-6-phosphate dehydrogenase (from Yeast)
J61181.4I
:
- Pk:1 KU
- CAS No.:9001-40-5
- Source:Yeast
- Storage conditions:–30...–10 °C
- Form:Lyophilized Powder
- MDL No.:MFCD00081656
- Enzyme name:Glucose-6-phosphate dehydrogenase
- Solubility:Soluble in water and dilute buffer
Applications: Converts p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+