Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Phosphate, 0,5M buffer solution, pH 6,5

Phosphate, 0,5M buffer solution, pH 6,5

Phosphate, 0,5M buffer solution, pH 6,5
Certificates
Phosphate, 0,5M buffer solution, pH 6,5

2 Options Available Below

Product Details & Documents

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J60845.AK
  • J60845.AP
  • Physical form:
  • Liquid
  • Liquid
  • N ° CAS.:
  • 7558-80-7
  • 7558-80-7
  • MDL No.:
  • MFCD00003527
  • MFCD00003527

Specifications

Product Family Options

Product Information

  • Désignation
    Concentration
    Valeur pH
    Cdt
    Availability
    Price
  • Phosphate buffer soln
    0,5 M
    6,5
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 104,00 €
    104,00 €
    1 * 250 mL
       104,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60845.AK
    Physical form
    Liquid
    N ° CAS.
    7558-80-7
    MDL No.
    MFCD00003527
  • Phosphate buffer soln
    0,5 M
    6,5
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 142,00 €
    142,00 €
    1 * 500 mL
       142,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60845.AP
    Physical form
    Liquid
    N ° CAS.
    7558-80-7
    MDL No.
    MFCD00003527
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.