Order Entry
Belgium
Orders LinkContactUsLinkComponent
SSC buffer (20X)

SSC buffer (20X)

SSC buffer (20X)
Certificates
SSC buffer (20X)

3 Options Available Below

Product Details & Documents

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00132371

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J60839.K2
  • J60839.K3
  • J60839.K7
  • Physical form:
  • Colourless liquid
  • Colourless liquid
  • Colourless liquid
  • MDL No.:
  • MFCD00132371
  • MFCD00132371
  • MFCD00132371

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • SSC buffer, 20X
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 55,70 €
    55,70 €
    1 * 1 L
       55,70 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60839.K2
    Physical form
    Colourless liquid
    MDL No.
    MFCD00132371
  • SSC buffer, 20X
    2 L
    Log in for availability
    Log in for availability
    1 * 2 L
    1 * 2 L - 85,80 €
    85,80 €
    1 * 2 L
       85,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60839.K3
    Physical form
    Colourless liquid
    MDL No.
    MFCD00132371
  • SSC buffer, 20X
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 156,00 €
    156,00 €
    1 * 4 L
       156,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60839.K7
    Physical form
    Colourless liquid
    MDL No.
    MFCD00132371
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.