CHAPS lysis buffer
Supplier: Thermo Fisher Scientific
About this item
For cell lysis and protein extraction.
Appearance: Clear liquid
MDL No.: MFCD00282866
Contains 50 mM Tris-HCl (pH 7,4), 110 mM NaCl, 5 mM EDTA and 1% CHAPS
2 Options Available Below
Product Details & Documents
For cell lysis and protein extraction.
Appearance: Clear liquid
MDL No.: MFCD00282866
Contains 50 mM Tris-HCl (pH 7,4), 110 mM NaCl, 5 mM EDTA and 1% CHAPS
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J60580.AD
- J60580.AE
- MDL No.:
- MFCD00282866
- MFCD00282866
- Appearance:
- Clear liquid
- Clear liquid
- Physical form:
- Liquid
- Liquid
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- J60580.ADCHAPS lysis bufferSpecifications:
More
Specifications
Cat. No.J60580.ADMDL No.MFCD00282866AppearanceClear liquidPhysical formLiquid - J60580.AECHAPS lysis bufferSpecifications:
More
Specifications
Cat. No.J60580.AEMDL No.MFCD00282866AppearanceClear liquidPhysical formLiquid
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



