Phosphate-buffered saline (PBS, 10X), no potassium
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
MDL No.: MFCD00131855
Specifications
- MDL No.:MFCD00131855
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together