TE buffer, pH 7,4, RNAse free
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.
Physical Form: Liquid
Appearance: Clear
MDL No.: MFCD00236359
- MDL No.:MFCD00236359
- Storage temperature:RT
- Physical form:Liquid
- Appearance:Clear
Frequently Bought Together