Order Entry
Belgium
ContactUsLinkComponent
TPE (10X), TRIS + phosphate + EDTA
TPE (10X), TRIS + phosphate + EDTA
Catalog # J60233.K2
CAS Number:  
TPE (10X), TRIS + phosphate + EDTA
Catalog # J60233.K2
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    TPE, 10X
  • MDL No.:
    MFCD00132488
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

Contains 0,9 M tris base, 0,9 M phosphate and 200 mM EDTA.

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00132488