Order Entry
Belgium
ContactUsLinkComponent
MOPS-EDTA-sodium acetate buffer (MESA)
MOPS-EDTA-sodium acetate buffer (MESA)
Certificates
MOPS-EDTA-sodium acetate buffer (MESA)
Certificates

Specifications

  • MDL No.:
    MFCD00131625
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together