Order Entry
Belgium
Orders LinkContactUsLinkComponent
POPSO, 0.2 M buffer solution, pH 8.0

POPSO, 0.2 M buffer solution, pH 8.0

POPSO buffer solution, 0,2 M, 8,0, 250 mL
Catalog # J60085.AK
POPSO, 0.2 M buffer solution, pH 8.0
Catalog # J60085.AK
CAS Number:  
POPSO, 0.2 M buffer solution, pH 8.0
POPSO, 0.2 M buffer solution, pH 8.0

POPSO buffer solution, 0,2 M, 8,0, 250 mL

CAS Number:  
Price
90,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 108321-07-9
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
MDL No.: MFCD00070005

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pk:
    250 mL
  • Description:
    POPSO buffer solution
  • pH value:
    8,0
  • Cat. No.:
    J60085.AK
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00070005
  • CAS No.:
    108321-07-9

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.