Order Entry
Belgium
Orders LinkContactUsLinkComponent
ADA, 0,2 M Buffer Solutions

ADA, 0,2 M Buffer Solutions

ADA, 0,2 M Buffer Solutions
Certificates
ADA, 0,2 M Buffer Solutions

5 Options Available Below

Product Details & Documents

N-(2-Acetimido)iminodiacetic acid monosodium salt (ADA) buffer solutions PH values 6,0 to 7,5.

CAS No.: 7415-22-7
Appearance: Colourless
MDL No.: MFCD00065483

Caution: Storage Temperature: 2 to 8°C

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J60054.AE
  • J61259.AE
  • J61259.AK
  • J62424.AE
  • J62424.AK
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • CAS No.:
  • 7415-22-7
  • 7415-22-7
  • 7415-22-7
  • 7415-22-7
  • 7415-22-7
  • MDL No.:
  • MFCD00065483
  • MFCD00065483
  • MFCD00065483
  • MFCD00065483
  • MFCD00065483
  • Appearance:
  • Colourless
  • Colourless
  • Colourless
  • Colourless
  • Colourless
  • Storage temperature:
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Concentration
    pH value
    Pack type
    Pk
    Availability
    Price
  • ADA buffer solution
    0,2 M
    6,0
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 55,50 €
    55,50 €
    1 * 100 mL
       55,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60054.AE
    Physical form
    Liquid
    CAS No.
    7415-22-7
    MDL No.
    MFCD00065483
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • ADA buffer solution
    0,2 M
    6,5
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 63,30 €
    63,30 €
    1 * 100 mL
       63,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61259.AE
    Physical form
    Liquid
    CAS No.
    7415-22-7
    MDL No.
    MFCD00065483
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • ADA buffer solution
    0,2 M
    6,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 76,80 €
    76,80 €
    1 * 250 mL
       76,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61259.AK
    Physical form
    Liquid
    CAS No.
    7415-22-7
    MDL No.
    MFCD00065483
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • ADA buffer solution
    0,2 M
    7,5
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 66,00 €
    66,00 €
    1 * 100 mL
       66,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62424.AE
    Physical form
    Liquid
    CAS No.
    7415-22-7
    MDL No.
    MFCD00065483
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • ADA buffer solution
    0,2 M
    7,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 79,80 €
    79,80 €
    1 * 250 mL
       79,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62424.AK
    Physical form
    Liquid
    CAS No.
    7415-22-7
    MDL No.
    MFCD00065483
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C