Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methylcyclohexane ≥99%

Methylcyclohexane ≥99%

Flammable
Health
Irritant
Environment
Methylcyclohexane ≥99%
Certificates
Methylcyclohexane ≥99%
Methylcyclohexane ≥99%

2 Options Available Below

Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,6047
  • Boiling Pt:101 °C
  • Melting Pt:–126 °C
  • Density:0.771
  • Flash Pt:–4 °C (–25 °F)
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Product Family Options

Product Information