L-Aspartic aciddiethyl ester hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16115-68-7
:
Formula:C₈H₁₅NO₄·HCl
MDL Number:MFCD00038910
:225.669 g/mol
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together