Order Entry
Belgium
ContactUsLinkComponent
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
Irritant
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
Catalog # H26897.03
CAS Number:  
Irritant
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride
Catalog # H26897.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:59831-02-6
 :
Formula:C₂₇H₂₆Cl₂P₂Pd
MDL Number:MFCD03844773
 :589.77 g/mol
 :GHS07
Storage Temperature:Ambient