Order Entry
Belgium
ContactUsLinkComponent
Ammonium dihydrogen phosphate 10% in nitric acid 2%, Specpure® matrix modifier solution
Corrosive
Ammonium dihydrogen phosphate 10% in nitric acid 2%, Specpure® matrix modifier solution
Catalog # 42663.AE
CAS Number:  
Corrosive
Ammonium dihydrogen phosphate 10% in nitric acid 2%, Specpure® matrix modifier solution
Catalog # 42663.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Melting Pt:2080 °C
Density:5.11 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :115.03 g/mol
EINECS:231-714-2
 :GHS05
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396