Phenylglyoxal monohydrate 97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 1075-06-5 Molecular Formula: C8H6O2.H2O Formula Weight: 152.15 (134.12anhy) Melting Point: 71-77° Boiling Point: 142°/125mm Beilstein Reference: 1854721 MDL No.: MFCD00149499
Synonyms: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone
- CAS Number:1075-06-5
- Formula:C₈H₆O₂·H₂O
- MDL Number:MFCD00149499
- Molecular Weight:152.15 g/mol (134.12 anhy)
- Boiling Pt:142 °C (125 mmHg)
- Melting Pt:71…77 °C
- Storage Temperature:Ambient
- EINECS:214-036-1
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 1075-06-5 Molecular Formula: C8H6O2.H2O Formula Weight: 152.15 (134.12anhy) Melting Point: 71-77° Boiling Point: 142°/125mm Beilstein Reference: 1854721 MDL No.: MFCD00149499
Synonyms: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone
- CAS Number:1075-06-5
- Formula:C₈H₆O₂·H₂O
- MDL Number:MFCD00149499
- Molecular Weight:152.15 g/mol (134.12 anhy)
- Boiling Pt:142 °C (125 mmHg)
- Melting Pt:71…77 °C
- Storage Temperature:Ambient
- EINECS:214-036-1
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



