Order Entry
Belgium
ContactUsLinkComponent
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L
Certificates
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L
Certificates

Specifications

  • CAS No.:
    9005-84-9
  • MDL No.:
    MFCD00082026
  • Physical form:
    Liquid, For iodometric titrations

Specifications

Related Information

Frequently Bought Together