Order Entry
Belgium
ContactUsLinkComponent
Yttrium, oil based standard solution, Y 1000 µg/g, Specpure®
Yttrium, oil based standard solution, Y 1000 µg/g, Specpure®
Catalog # 47172.18
CAS Number:  
Yttrium, oil based standard solution, Y 1000 µg/g, Specpure®
Catalog # 47172.18
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    oil
  • Pk:
    50 g
  • MDL No.:
    MFCD00011468
  • Element:
    Yttrium
  • Symbol:
    Y

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00011468