Order Entry
Belgium
ContactUsLinkComponent
Cobalt, oil based standard solution, Co 1000 µg/g, Specpure®
Cobalt, oil based standard solution, Co 1000 µg/g, Specpure®
Catalog # 47143.18
CAS Number:  
Cobalt, oil based standard solution, Co 1000 µg/g, Specpure®
Catalog # 47143.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Mineral oil
  • Pk:
    50 g
  • MDL No.:
    MFCD00010935
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00010935