Tellurium, AAS standard solution, Te 1000 µg/ml, Specpure®
Catalog # 47130.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:30% HCl
- Pk:100 mL
- Environmentally Preferable:
- MDL No.:MFCD00134062
- Element:Tellurium
- Symbol:Te